Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M8309

Sigma-Aldrich

[Ala113]-Myelin Basic Protein Fragment 104-118

synthetic, lyophilized powder

Synonym(s):

[Ala113]-MBP fragment 104-118

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

biological source

synthetic

form

lyophilized powder

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)CNC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC(=O)[C@H](CCCCN)NC(=O)CN)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](CO)C(=O)N[C@@H](Cc2c[nH]c3ccccc23)C(=O)NCC(=O)N[C@@H](C)C(O)=O

InChI

1S/C67H104N20O19/c1-35(2)23-45(81-55(94)31-75-58(97)44(20-14-22-72-67(70)71)80-54(93)30-74-57(96)43(19-12-13-21-68)79-52(91)27-69)60(99)86-50(33-89)64(103)83-46(24-36(3)4)61(100)85-49(32-88)63(102)78-37(5)56(95)82-47(25-39-15-8-7-9-16-39)62(101)87-51(34-90)65(104)84-48(26-40-28-73-42-18-11-10-17-41(40)42)59(98)76-29-53(92)77-38(6)66(105)106/h7-11,15-18,28,35-38,43-51,73,88-90H,12-14,19-27,29-34,68-69H2,1-6H3,(H,74,96)(H,75,97)(H,76,98)(H,77,92)(H,78,102)(H,79,91)(H,80,93)(H,81,94)(H,82,95)(H,83,103)(H,84,104)(H,85,100)(H,86,99)(H,87,101)(H,105,106)(H4,70,71,72)/t37-,38-,43-,44-,45-,46-,47-,48-,49-,50-,51-/m0/s1

InChI key

FBVMKAAYUXGGSU-NBRSAOMZSA-N

Gene Information

human ... MBP(4155)

Amino Acid Sequence

Gly-Lys-Gly-Arg-Gly-Leu-Ser-Leu-Ser-Ala-Phe-Ser-Trp-Gly-Ala

Biochem/physiol Actions

Inhibitor of Protein Kinase C

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H D Su et al.
Biochemical and biophysical research communications, 134(1), 78-84 (1986-01-14)
Synthetic peptide analogs of the bovine myelin basic protein (MBP) corresponding to residues 104-118 were found to specifically inhibit phospholipid/ Ca2+-dependent protein kinase (protein kinase C). The peptides [Ala107]MBP (104-118) and [Ala113]MBP (104-118) inhibited protein phosphorylation of intact MBP, histone

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service