M7556
Anti-MAP Kinase 2 (ERK-2) antibody produced in rabbit
IgG fraction of antiserum, buffered aqueous solution, ~1 mg/mL
Synonym(s):
Anti-ERK-2
About This Item
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 42 kDa
species reactivity
rat, human, mouse
concentration
~1 mg/mL
technique(s)
immunoprecipitation (IP): 5 μg using 0.5-1 mg of a mouse 3T3 fibroblast lysate
western blot: 0.5-2 μg/mL using human A431 cells, mouse 3T3 fibroblasts or rat L6 cells
UniProt accession no.
shipped in
dry ice
Storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MAPK1(5594)
mouse ... Ephb2(13844) , Mapk1(26413)
rat ... Mapk1(116590)
General description
Anti-MAP Kinase 2 (ERK-2) antibody is specific for MAPK2 (42 kDa) in mice, humans and rats. This antibody does not cross react with MAP kinase 1 (ERK-1) nor does it interfere with the kinase activity.
Immunogen
application
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service