Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6569

Sigma-Aldrich

Anti-MBD1 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~80 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

indirect immunofluorescence: 1-2 μg/mL using HeK 293T cells transfected with MBD1 (isoform 3), fixed with paraformaldehyde-triton
microarray: suitable
western blot: 0.25-0.5 μg/mL using extracts of HEK 293T cells transfected with MBD1 (isoform 3)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MBD1(4152)

General description

MBD1 gene is mapped to human chromosome 18q21.1.
Methyl-CpG binding domain protein 1 (MBD1) exists as five isoforms.

Specificity

Anti MBD1 recognizes recombinant human MBD1 (~80 kDa).

Immunogen

synthetic peptide corresponding to amino acids 591-605 of human MBD1, conjugated to KLH via an N-terminal added lysine residue. This sequence is found in MBD1 isoforms.

Application

Anti-MBD1 antibody produced in rabbit may be used for immunofluorescence and immunoblotting.

Biochem/physiol Actions

MBD1 can act as a transcriptional repressor. It mediates transcriptional repression by recruiting heterochromatin-binding protein (HP1), the histone methyl transferase Suv39h1 and histone deacetylase using the methyl binding domain (MBD). The transcriptional repression domain (TRD) of MBD1 mediates its interaction with MBD1-containing chromatin-associated factor (MCAF-1).
Methyl-CpG binding domain protein 1 (MBD1) aids pancreatic cancer cells in drug resistance and contributes to disease progression. It gets recruited to those sites where DNA damage has occurred and the protein has also shown to be involved in suppression of gene transcription. MBD1 acts as a link between methylation of DNA and repression of transcription.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in frost-free freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilu-tions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silencing of MBD1 reverses pancreatic cancer therapy resistance through inhibition of DNA damage repair.
Xu J
International Journal of Oncology, 42(6), 2046-2052 (2013)
Takaya Ichimura et al.
The Journal of biological chemistry, 280(14), 13928-13935 (2005-02-05)
DNA methylation cooperates with methylation at lysine 9 of histone H3 (H3-K9), a modified histone molecule that is targeted by heterochromatin protein 1, to form a transcriptionally silent chromatin. Methyl CpG-binding protein MBD1 recognizes methylated CpG dinucleotide and recruits H3-K9
Holly N Cukier et al.
Neurogenetics, 11(3), 291-303 (2009-11-19)
Misregulation of the methyl-CpG-binding protein 2 (MECP2) gene has been found to cause a myriad of neurological disorders including autism, mental retardation, seizures, learning disabilities, and Rett syndrome. We hypothesized that mutations in other members of the methyl-CpG-binding domain (MBD)
Proteomic analysis of differential proteins in pancreatic carcinomas: Effects of MBD1 knock-down by stable RNA interference.
Liu C
BMC Cancer, 8, 121-121 (2008)
Mechanism of transcriptional regulation by methyl-CpG binding protein MBD1.
Fujita N
Molecular and Cellular Biology, 20(14), 5107-5118 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service