Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6567

Sigma-Aldrich

Monoclonal Anti-MRP3 antibody produced in mouse

250 μg/mL, clone M3II-21, tissue culture supernatant

Synonym(s):

Anti-Multidrug Resistance Associated Protein 3

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

M3II-21, monoclonal

mol wt

antigen 190-200 kDa

species reactivity

human

concentration

250 μg/mL

technique(s)

immunocytochemistry: 1:20-1:50 using acetone- or formaldehyde-fixed cytospin preparations
immunohistochemistry (frozen sections): 1:20 using acetone-fixed sections
western blot: 1:20-1:50

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ABCC3(8714)

General description

Multidrug Resistance-Associated Protein (MRP) is a seven member subfamily of ABC (ATP-binding cassette) transporters. MRP1, 2, and 3 have a similar structure with the ability to transport glutathione and glucuronate conjugates. MRP4 and MRP5 share more structure similarity with each other than with MRP1, 2, and 3. MRP4 and MRP5 also have the ability to transport cyclic nucleotides..The monoclonal anti-MRP3 recognizes human Multidrug Resistance-Associated Protein 3 (MRP3, 190-200 kDA)..

Specificity

Reacts with an internal epitope of MRP3. Does not cross-react with the human MDR1, MRP1, MRP2, or MRP5 gene products.

Immunogen

fusion protein of MRP3 (amino acids 830-949).

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
The antibody may be used in immunocytochemistry, immunoblotting, immunohistochemistry.

Physical form

Supplied in serum-free medium containing 0.7% bovine serum albumin and 0.1% sodium azide.

Preparation Note

Monoclonal Anti-Multidrug Resistance-Associated Protein 3 (MRP3), mouse isotype IgG1, derived using M3II-21 hybridoma produced by fusion SP2/O mouse myeloma cells and Balb/c mouse lymph nodes cells immunized with amino acids 830-949 of the human MRP3 bacterial fusion protein. The antibody is concentrated from culture supernatant of M3II-21 hybridoma cells grown in a bioreactor.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junro Kataoka et al.
Oncology reports, 35(5), 2576-2582 (2016-03-18)
Runt-related transcription factor 3 (RUNX3) is known to function as a tumor suppressor in gastric cancer and other types of cancers, including hepatocellular carcinoma (HCC). However, its role has not been fully elucidated. In the present study, we aimed to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service