Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6542

Sigma-Aldrich

Macrophage Inflammatory Protein-1β from mouse

recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

mMIP-1β, MIP-1β

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202

biological source

mouse

recombinant

expressed in E. coli

assay

≥97% (SDS-PAGE and N-terminal analysis)

form

lyophilized powder

potency

40-100 ng/mL ED50/EC50

mol wt

7.8 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

mouse ... Ccl4(20303)

General description

MIP-1α and MIP-1β (macrophage inflammatory protein) were originally co-purified from LPS stimulated mouse macrophages. Recombinant, mouse MIP-1β (rmMIP-1β) consists of 69 amino acids. MIP- 1β belongs to the chemokine β subfamily which is characterized by a C-C configuration at the first two cysteines. MIP-1β has endogenous pyrogenic activity when it is injected intravenously into rabbits. Although other cytokines, such as IL-1alpha, IL-1β and TNF have endogenous pyrogenic activity, the pyrogenic effects of these cytokines can be inhibited by cyclooxgenase blockers, while the pyrogenicity of MIP- 1β is unaffected by these agents. MIP-1β can synergize with the hematopoietic growth factors granulocyte macrophage CSF (GM-CSF) or macrophage CSF (M-CSF) to enhance colony formation.

Biochem/physiol Actions

MIP-1β has endogenous pyrogenic activity when it is injected intravenously into rabbits. MIP-1 can synergize with the hematopoietic growth factors granulocytemacrophage CSF (GM-CSF) or macrophage CSF (M-CSF) to enhance colony formation.

Physical form

Lyophilized from a 0.2 μm filtered solution in 30% acetonitrile and 0.1% trifluoroacetic acid containing 0.5 mg bovine serum albumin

Analysis Note

The biological activity of MIP-1β was tested in culture by measuring its ability to inhibit hematopoietic stem cell proliferation in an in vitro colony assay.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G J Graham et al.
Nature, 344(6265), 442-444 (1990-03-29)
The haemopoietic system has three main compartments: multi-potential stem cells, intermediate stage progenitor cells and mature cells. The availability of simple reproducible culture systems has made possible the characterization and purification of regulators of the progenitor cells, including colony-stimulating factors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service