Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6525

Sigma-Aldrich

D-Mannose Agarose

saline suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41106500

form

saline suspension

matrix

Cross-linked 4% beaded agarose

matrix activation

epoxy

matrix attachment

hydroxyl

matrix spacer

12 atoms

capacity

≥40 mg/mL binding capacity (Concanavalin A)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Physical form

Suspension in 0.5 M NaCl containing preservative

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chanez Chemani et al.
Infection and immunity, 77(5), 2065-2075 (2009-02-25)
Pseudomonas aeruginosa is a frequently encountered pathogen that is involved in acute and chronic lung infections. Lectin-mediated bacterium-cell recognition and adhesion are critical steps in initiating P. aeruginosa pathogenesis. This study was designed to evaluate the contributions of LecA and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service