Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6417

Sigma-Aldrich

Macrophage Inflammatory Protein-1β human

≥97% (SDS-PAGE and N-terminal analysis), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture

Synonym(s):

MIP-1β

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

human

recombinant

expressed in baculovirus infected Sf21 cells

assay

≥97% (SDS-PAGE and N-terminal analysis)

form

lyophilized powder

potency

10-30 ng/mL ED50 (for monocytes)
3-10 ng/mL ED50 (for mouse BAF/3 cells transfected with HCCR5 cells)

mol wt

7.8 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... CCL3(6348)

Physical form

Lyophilized from a 0.2 μm filtered solution of 30% acetonitrile and 0.1% trifluoroacetic acid containing 0.5 mg bovine serum albumin.

Analysis Note

The biological activity is measured by its ability to chemoattract cultured human monocytes or mouse BaF/3 cells transfected with human CCR5.

Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service