Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6315

Sigma-Aldrich

Monoclonal Anti-Mismatch Repair Protein 2 antibody produced in mouse

~1 mg/mL, clone 2MSH01, tissue culture supernatant

Synonym(s):

Anti-MSH2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

tissue culture supernatant

antibody product type

primary antibodies

clone

2MSH01, monoclonal

mol wt

antigen 102 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MSH2(4436)

General description

Mismatch repair protein 2 (MSH2) possesses four domains and belongs to the mismatch repair (MMR) gene family. The gene encoding this protein is localized on human chromosome 2p21.

Immunogen

recombinant human mismatch repair protein 2 (MSH2).

Biochem/physiol Actions

Mismatch repair protein 2 (MSH2) plays a very important role in the DNA mismatch repair system. It repairs DNA damage which is induced by drugs. MSH2 dysfunction has been associated with nonpolyposis colorectal cancer (Lynch syndrome).

Physical form

Contains 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Chen et al.
Scandinavian journal of immunology, 81(2), 121-128 (2014-11-21)
γδT cells function as sentinels in early host responses to infections and malignancies. Previously, we found ectopically expressed human MutS homologue 2 (hMSH2), recognized by γδT cells, triggered a γδT cell-mediated cytolysis to tumor cells. However, the characteristics of hMSH2-specific
Jin Zhang et al.
Oncology reports, 32(5), 2199-2206 (2014-09-02)
The objective of the present study was to investigate the association between paclitaxel resistance, gene copy number, and gene expression in ovarian carcinoma, and to restore paclitaxel sensitivity in a paclitaxel-resistant ovarian carcinoma cell line by using hMSH2-targeting siRNA. Paclitaxel-resistant
Jen-Chung Ko et al.
Biochemical and biophysical research communications, 456(1), 506-512 (2014-12-10)
The anti-estrogen tamoxifen has been used worldwide as an adjuvant hormone therapeutic agent in the treatment of breast cancer. However, the molecular mechanism of tamoxifen-induced cytotoxicity in non-small cell lung cancer (NSCLC) cells has not been identified. Human MutS homolog

Articles

DNA damage and repair mechanism is vital for maintaining DNA integrity. Damage to cellular DNA is involved in mutagenesis, the development of cancer among others.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service