Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M6184

Sigma-Aldrich

Anti-Matrix Metalloproteinase-23, C-Terminal antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MMP-23

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen (active form) 45 kDa
antigen (pro-form) 58 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:1,000

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP23B(8510)

General description

Matrix metalloproteinase-23B (MMP-23B) possesses regions which are rich in amino acids like proline and cysteine. The MMP-23B gene is localized on human chromosome 1p36.

Specificity

Reacts with reduced and non-reduced MMP-23. Recognizes the pro-form and active forms of MMP-23 as well as a series of further cleaved active forms. The hemopexin domain is removed from the smallest active form of MMP-23 and the antibody does not detect this form.

Immunogen

synthetic peptide corresponding to the C-terminal of human matrix metalloproteinase-23 (MIFR-2, CA-MMP).

Biochem/physiol Actions

Matrix metalloproteinase-23B (MMP-23B) is part of the matrix metalloproteinase family of proteins, which take part in the remodeling of the extracellular matrix.

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriela Fonseca-Camarillo et al.
Scandinavian journal of immunology, 93(1), e12962-e12962 (2020-08-28)
It has been reported that EMMPRIN is involved in the regulation of immune response and the induction of MMPs production by fibroblasts. The aim of this study was to describe the intestinal gene expression and protein production of EMMPRIN, MMP23
Marzena Gajecka et al.
European journal of human genetics : EJHG, 13(2), 139-149 (2004-10-16)
Structural chromosome abnormalities have aided in gene identification for over three decades. Delineation of the deletion sizes and rearrangements allows for phenotype/genotype correlations and ultimately assists in gene identification. In this report, we have delineated the precise rearrangements in four
Matrix metalloproteinases.
H Nagase et al.
The Journal of biological chemistry, 274(31), 21491-21494 (1999-07-27)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service