Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5823

Sigma-Aldrich

Anti-MIB2 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Mindbomb homolog 2, Anti-Novel zinc finger protein, Anti-Novelzin, Anti-Skeletrophin, Anti-Zinc finger ZZ type with ankyrin repeat domain protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~110 kDa

species reactivity

human, mouse

technique(s)

immunoprecipitation (IP): 2-5 μL using NIH3T3 cell lysate
western blot: 1:500-1:1,000 using lysates of NIH3T3 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... MIB2(142678)
mouse ... Mib2(76580)

General description

Mindbomb homolog 2 (MIB2), which is also known as skeletrophin, is an ubiquitin ligase. The gene encoding it is localized on human chromosome 1p36.32.

Immunogen

synthetic peptide corresponding to amino acids 507-524 of human MIB2 conjugated to KLH. The corresponding sequence is identical in mouse.

Biochem/physiol Actions

Mindbomb homolog 2 (MIB2) takes part in the ubiquitination of Notch ligands like Jagged-2 (JAG2). It binds to actin and CYLD (cylindromatosis (turban tumor syndrome)), which is an ubiquitin hydrolase. It has also been shown that MIB2 negatively regulates the transcription of Met oncogene.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Takeuchi et al.
Oncogene, 25(53), 7059-7069 (2006-05-23)
Skeletrophin (mindbomb homolog 2 (MIB2)) is a RING (Really Interesting New Gene) finger-dependent ubiquitin ligase, which targets the intracellular region of Notch ligands. A previous immunohistochemical study demonstrated that skeletrophin was downregulated in many melanomas. In the present study, we
Tamotsu Takeuchi et al.
DNA and cell biology, 24(5), 339-344 (2005-05-05)
Recent experiments have revealed that aberrant functionality of the chromatin remodeling complex is related to tumorigenicity in various malignant tumors. Skeletrophin is an actin-binding cytoskeleton-related molecule, which is induced by the overexpression of truncated human SWI1 (SMARCF1). Human SWI1 is
Neil Rajan et al.
Oncotarget, 5(23), 12126-12140 (2015-01-08)
CYLD, an ubiquitin hydrolase, has an expanding repertoire of regulatory roles in cell signalling and is dysregulated in a number of cancers. To dissect CYLD function we used a proteomics approach to identify CYLD interacting proteins and identified MIB2, an

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service