Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5813

Sigma-Aldrich

Mimetic Orange 2 Ligand Affinity Adsorbent

suspension

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817

form

suspension

shipped in

wet ice

storage temp.

2-8°C

Application

Ligand affinity adsorbents are used in protein chromatography, affinity chromatography and dye resins. Ligand affinity adsorbents have been used to describe a simple protocol for the purification of Taq Polymerase from E. coli lysates as well as for the purification of other DNA polymerases.

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

95.0 °F - closed cup

flash_point_c

35 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huma Khan et al.
The FEBS journal, 272(18), 4660-4671 (2005-09-15)
The roles of His181, His184 and Tyr186 in PETN reductase have been examined by mutagenesis, spectroscopic and stopped-flow kinetics, and by determination of crystallographic structures for the Y186F PETN reductase and reduced wild-type enzyme-progesterone complex. Residues His181 and His184 are
Crystal structure of pentaerythritol tetranitrate reductase: "flipped" binding geometries for steroid substrates in different redox states of the enzyme.
Barna, T.M., et al.
Journal of Molecular Biology, 310(2), 443-447 (2001)
Biological activity and biosynthesis of pentacyclic oxylipins: The linoleic acid pathway.
Gundlach, H., and Zenk, M.H.
Phytochemistry, 47(4), 527-537 (1998)
Huma Khan et al.
The Journal of biological chemistry, 279(29), 30563-30572 (2004-05-07)
The structure of pentaerythritol tetranitrate (PETN) reductase in complex with the nitroaromatic substrate picric acid determined previously at 1.55 A resolution indicated additional electron density between the indole ring of residue Trp-102 and the nitro group at C-6 of picrate.
Huma Khan et al.
The Journal of biological chemistry, 277(24), 21906-21912 (2002-03-30)
The reaction of pentaerythritol tetranitrate reductase with reducing and oxidizing substrates has been studied by stopped-flow spectrophotometry, redox potentiometry, and X-ray crystallography. We show in the reductive half-reaction of pentaerythritol tetranitrate (PETN) reductase that NADPH binds to form an enzyme-NADPH

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service