Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5808

Sigma-Aldrich

Anti-Matrix Metalloproteinase-14, N-Terminal antibody produced in rabbit

0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-MMP-14

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~65 kDa

species reactivity

human

concentration

0.5 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100 using human cancer tissue
indirect ELISA: 1:1,000
western blot: 1:100-1:500 using A375 lysate

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP14(4323)

General description

Matrix metalloproteinase-14 (MMP-14) is a collagenase localized to the cell membrane. The gene encoding it is present on human chromosome 14.

Immunogen

synthetic peptide corresponding to amino acids 185-299 in the N-terminal region of human matrix metalloproteinase-14 (membrane-type matrix metalloproteinase-1), conjugated to KLH.

Application

Anti-matrix metalloproteinase-14, N-terminal antibody produced in rabbit has been used for western blotting at a dilution of 1:100.

Biochem/physiol Actions

Matrix metalloproteinase-14 (MMP-14) aids in the migration of leukocytes and breaks down collagen. Migration of cells through the extracellular matrix is also mediated by it. MMP-14 has been shown to be associated with tuberculosis and glioma.

Physical form

Solution in phosphate buffered saline containing 0.09% (w/v) sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V M Kähäri et al.
Experimental dermatology, 6(5), 199-213 (1998-02-05)
Matrix metalloproteinases (MMPs) are a family of zinc-dependent endopeptidases collectively capable of degrading essentially all extracellular matrix components. These enzymes can be produced by several different types of cells in skin such as fibroblasts, keratinocytes, macrophages, endothelial cells, mast cells
Takao Ide et al.
International journal of cancer, 119(12), 2750-2759 (2006-09-26)
The hypoxic environment in tumor is reported to play an important role in pancreatic cancer progression. The interaction between stromal and cancer cells also contributes to the malignant behavior of pancreatic cancer. In the present study, we investigated whether hypoxic
Tarangini Sathyamoorthy et al.
Journal of immunology (Baltimore, Md. : 1950), 195(3), 882-891 (2015-06-21)
Tuberculosis (TB) remains a global pandemic and drug resistance is rising. Multicellular granuloma formation is the pathological hallmark of Mycobacterium tuberculosis infection. The membrane type 1 matrix metalloproteinase (MT1-MMP or MMP-14) is a collagenase that is key in leukocyte migration
Piero Pingitore et al.
Human molecular genetics, 25(23), 5212-5222 (2016-10-16)
Liver fibrosis is a pathological scarring response to chronic hepatocellular injury and hepatic stellate cells (HSCs) are key players in this process. PNPLA3 I148M is a common variant robustly associated with liver fibrosis but the mechanisms underlying this association are
Ilya Ulasov et al.
Cancer letters, 365(2), 240-250 (2015-06-09)
MMP14 (MT1-MMP) is a cell membrane-associated proteinase of the extracellular matrix, whose biological roles vary from angiogenesis to cell proliferation and survival. We recently found a direct correlation between MMP14 expression levels in brain tumors of glioma patients and the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service