Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M5781

Sigma-Aldrich

18-Methylnonadecanoic acid

≥98% (capillary GC)

Synonym(s):

Isoarachidic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H40O2
CAS Number:
Molecular Weight:
312.53
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

assay

≥98% (capillary GC)

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

2-8°C

SMILES string

CC(C)CCCCCCCCCCCCCCCCC(O)=O

InChI

1S/C20H40O2/c1-19(2)17-15-13-11-9-7-5-3-4-6-8-10-12-14-16-18-20(21)22/h19H,3-18H2,1-2H3,(H,21,22)

InChI key

UCDAVJCKGYOYNI-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Kaneda
Microbiological reviews, 55(2), 288-302 (1991-06-01)
Branched-chain fatty acids of the iso and anteiso series occur in many bacteria as the major acyl constituents of membrane lipids. In addition, omega-cyclohexyl and omega-cycloheptyl fatty acids are present in several bacterial species. These two types of fatty acids
Róbert Kubinec et al.
Journal of chromatography. A, 1218(13), 1767-1774 (2011-02-26)
Isomer mixtures of monomethyl branched saturated C7-C23 fatty acid methyl esters (FAME) were prepared by performing a methylene insertion reaction to the straight chain FAME and this study model was completed by using commercially available standards of C4-C7 FAME. The
Jeroen S Dickschat et al.
Organic & biomolecular chemistry, 3(15), 2824-2831 (2005-07-21)
The fatty acid (FA) profiles of the myxobacteria Stigmatella aurantiaca and Myxococcus xanthus were investigated by acidic methanolysis of total cell extracts and GC or GC-MS analysis. The main components were 13-methyltetradecanoic acid (iso-15:0) and (Z)-hexadec-11-enoic acid (16:1, omega-5 cis).

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service