Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5560

Sigma-Aldrich

(S)-2-Amino-2-methyl-4-phosphonobutyric acid hydrochloride

synthetic, solid

Synonym(s):

(S)-MAP4 hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

biological source

synthetic

Quality Level

form

solid

color

white

SMILES string

Cl.C[C@](N)(CCP(O)(O)=O)C(O)=O

InChI

1S/C5H12NO5P.ClH/c1-5(6,4(7)8)2-3-12(9,10)11;/h2-3,6H2,1H3,(H,7,8)(H2,9,10,11);1H/t5-;/m0./s1

InChI key

DNFXUQUECNSSJF-JEDNCBNOSA-N

Gene Information

Biochem/physiol Actions

Selective mGluR4,6,7 metabotropic glutamate receptor antagonist.

Features and Benefits

This compound is featured on the Glutamate Receptors (G Protein Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T E Salt et al.
Neuroscience, 65(1), 5-13 (1995-03-01)
A variety of metabotropic excitatory amino acid receptors are present in the thalamus. We have investigated the possibility that some of these receptors may have presynaptic effects on GABAergic inhibitory transmission in the thalamus. Inhibitory responses in ventrobasal thalamic neurons
T Knöpfel et al.
Neuropharmacology, 34(8), 1099-1102 (1995-08-01)
The two reported metabotropic glutamate receptor (mGluR) antagonists, alpha-methyl-cyclopropyl glycine (MCCG) and alpha-methyl-aminophosphonobutyrate (MAP4) were tested on the mGluR1b, mGluR2 and mGluR4a subtypes of human mGluRs. Neither MCCG (500 microM) nor MAP4 (500 microM) antagonized the activation of mGluR1b by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service