Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5442

Sigma-Aldrich

Anti-Matrix Metalloproteinase-26, Hinge Region antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MMP-26

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MMP26(56547)

General description

Matrix metalloproteinase-26 (MMP-26) is also known as endometase and matrilysin-2. It is expressed in normal tissues and also in human carcinoma cells. MMP-26 possesses a propeptide domain, a signal peptide and a catalytic domain but does not have the hemopexin-like domain which is common to other members of its family. The MMP-26 gene is localized to human chromosome 11p15.3.

Specificity

By immunoblotting against the reduced protein, the antibody identifies bands at 30 kDa and 18 kDa (zymogen and active MMP-26, respectively).

Immunogen

synthetic peptide corresponding to the hinge region of human matrix metalloproteinase-26.

Biochem/physiol Actions

Matrix metalloproteinase-26 (MMP-26) degrades fibronectin, type IV collagen and activated pro-metalloproteinase-9. MMP-26 also aids in development of tumors.

Physical form

Solution in 0.01 M phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qinglin Hu et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(4), 3569-3574 (2013-12-10)
Tumor recurrence and metastasis are pressing issues of patients with colorectal cancer who receive surgery. Matrilysin-2 (MMP-26) has been proved to play an important role during invasion and metastasis of some human solid tumor. We aimed to investigate the clinical
Hirotaka Nishi et al.
Oncology reports, 30(2), 751-756 (2013-06-12)
The human matrix metalloproteinase (MMP)-26, also called matrilysin-2 or endometase, has been isolated as a matrilysin (MMP-7) homolog. Several reports describe that MMP-26 may be related to the development of endometrial carcinomas. Total RNAs were isolated from 51 normal endometrial
Zahraa I Khamis et al.
Journal of Cancer, 4(4), 296-303 (2013-04-10)
Human endometase/matrilysin-2/matrix metalloproteinase-26 (MMP-26) is an endopeptidase mostly produced by human carcinoma cells. While MMPs are thought to regulate the dynamics of extracellular matrix turnover, new evidence shows that these enzymes may play a critical regulatory role in inflammation. To
Yang Zhang et al.
Molecular medicine reports, 4(6), 1201-1209 (2011-08-02)
Matrix metalloproteinase 26 (MMP-26) is a novel member of the matrix metalloproteinase (MMP) family and is widely expressed in cancer cells of epithelial origin. MMP-26 has been shown to contribute to tumor development and to the restoration of tissue injury.
Helen E Gruber et al.
Experimental and molecular pathology, 92(1), 59-63 (2011-09-29)
Matrix metalloproteinase (MMP) regulation and expression is important in the aging/degenerating human intervertebral disc. MMP-26 (also known as matrilysin-2 or endometase) is a newly discovered MMP which degrades type IV collagen, fibronectin, fibrinogen, vitronectin, denatured collagen types I-IV, insulin-like growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service