Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5434

Sigma-Aldrich

Anti-Matrix Metalloproteinase-19, Hinge Region antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MMP-19

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen (active form) 45 kDa
antigen (pro-form) 58 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunohistochemistry (frozen sections): suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MMP19(4327)

General description

Matrix metalloproteinase-19 (MMP19) is an endopeptidase which depends on zinc for activation. It contains a signal sequence, a propeptide domain, a catalytic domain and a carboxy-terminal fragment. The gene encoding this protein is located on human chromosome 12q14.

Specificity

Reacts with reduced and non-reduced MMP-19. Recognizes the pro-form and active forms of MMP-19, as well as a series of further cleaved active forms.

Immunogen

synthetic peptide corresponding to the hinge region of human matrix metalloproteinase-19 (RASI).

Biochem/physiol Actions

Matrix metalloproteinase-19 (MMP-19) may have a role in arthritis and takes part in the advancement of lung cancer.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J O Stracke et al.
The Journal of biological chemistry, 275(20), 14809-14816 (2000-05-16)
We have recently cloned MMP-19, a novel matrix metalloproteinase, which, due to unique structural features, was proposed to represent the first member of a new MMP subfamily (Pendás, A. M., Knäuper, V. , Puente, X. S., Llano, E., Mattei, M.
Guoying Yu et al.
American journal of respiratory and critical care medicine, 190(7), 780-790 (2014-09-25)
Lung cancer is the leading cause of cancer death in both men and women in the United States and worldwide. Matrix metalloproteinases (MMPs) have been implicated in the development and progression of lung cancer, but their role in the molecular
J O Stracke et al.
FEBS letters, 478(1-2), 52-56 (2000-08-03)
Matrix metalloproteinase (MMP)-19 and MMP-20 (enamelysin) are two recently discovered members of the MMP family. These enzymes are involved in the degradation of the various components of the extracellular matrix (ECM) during development, haemostasis and pathological conditions. Whereas MMP-19 mRNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service