Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5318

Sigma-Aldrich

mC2p-NLS

solid

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C155H247N47O35
Molecular Weight:
3328.91
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

UniProt accession no.

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](Cc1c[nH]cn1)NC(=O)[C@H](C)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](NC(=O)[C@@H]2CCCN2C(=O)[C@@H](NC(=O)CNC(=O)[C@H](Cc3ccccc3)NC(=O)[C@H](C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](C)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCCN)NC(=O)[C@H](CCCCN)NC(=O)[C@@H]4CCCN4)C(C)C)C(C)C)[C@@H](C)O)C(C)C)C(C)C)[C@@H](C)O)C(=O)N[C@@H](Cc5c[nH]c6ccccc56)C(=O)N[C@@H](Cc7ccc(O)cc7)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](C)C(O)=O

InChI

1S/C146H238N46O33/c1-75(2)66-103(131(213)184-106(69-88-71-165-92-39-21-20-38-91(88)92)133(215)183-105(68-87-50-52-90(195)53-51-87)132(214)178-99(46-32-62-162-144(153)154)124(206)171-83(14)142(224)225)185-140(222)116(85(16)194)191-138(220)113(78(7)8)189-137(219)112(77(5)6)187-130(212)102(54-55-110(197)198)179-134(216)107(70-89-72-159-74-167-89)182-119(201)82(13)170-139(221)115(84(15)193)190-129(211)101(48-34-64-164-146(157)158)180-136(218)111(76(3)4)188-135(217)108-49-35-65-192(108)141(223)114(79(9)10)186-109(196)73-166-120(202)104(67-86-36-18-17-19-37-86)181-118(200)81(12)169-123(205)98(45-31-61-161-143(151)152)172-117(199)80(11)168-122(204)94(40-22-26-56-147)174-128(210)100(47-33-63-163-145(155)156)177-127(209)97(43-25-29-59-150)176-126(208)96(42-24-28-58-149)175-125(207)95(41-23-27-57-148)173-121(203)93-44-30-60-160-93/h17-21,36-39,50-53,71-72,74-85,93-108,111-116,160,165,193-195H,22-35,40-49,54-70,73,147-150H2,1-16H3,(H,159,167)(H,166,202)(H,168,204)(H,169,205)(H,170,221)(H,171,206)(H,172,199)(H,173,203)(H,174,210)(H,175,207)(H,176,208)(H,177,209)(H,178,214)(H,179,216)(H,180,218)(H,181,200)(H,182,201)(H,183,215)(H,184,213)(H,185,222)(H,186,196)(H,187,212)(H,188,217)(H,189,219)(H,190,211)(H,191,220)(H,197,198)(H,224,225)(H4,151,152,161)(H4,153,154,162)(H4,155,156,163)(H4,157,158,164)/t80-,81-,82-,83-,84+,85+,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,111-,112-,113-,114-,115-,116-/m0/s1

InChI key

GYXSOMLASXTNHL-BWYVTNJHSA-N

Gene Information

human ... CDK2(1017)

Amino Acid Sequence

Pro-Lys-Lys-Lys-Arg-Lys-Ala-Arg-Ala-Phe-Gly-Val-Pro-Val-Arg-Thr-Ala-His-Glu-Val-Val-Thr-Leu-Trp-Tyr-Arg-Ala

Biochem/physiol Actions

Peptide that contains the nuclear localization sequences (NLS); can be delivered intracellularly using liposomes for cell based studies and for its effect on HIV replication.
mC2p-NLS is a peptide that contains the nuclear localization sequences (NLS) and can be delivered intracellularly using liposomes for cell based studies and for its effect on HIV replication.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service