Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5272

Sigma-Aldrich

Anti-Mouse IgM (μ-chain specific) antibody produced in goat

IgG fraction of antiserum, lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

goat

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

secondary antibodies

clone

polyclonal

form

lyophilized powder

technique(s)

Ouchterlony double diffusion: 1:8

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Reconstitution

Reconstitute with 2 ml deionized water.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Christensen et al.
Applied and environmental microbiology, 58(4), 1244-1248 (1992-04-01)
Immunomagnetic beads (IMB) were used to recover thermophilic sulfate-reducing bacteria from oil field waters from oil production platforms in the Norwegian sector of the North Sea. IMB coated with polyclonal antibodies against whole-cell antigens of the thermophilic Thermodesulfobacterium mobile captured

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service