M5167
Myelin Basic Protein Guinea Pig Fragment 68-82
≥97% (HPLC)
Synonym(s):
des-Gly77, des-His78-Myelin Basic Protein Bovine fragment 68-84
About This Item
biological source
guinea pig
assay
≥97% (HPLC)
form
lyophilized
UniProt accession no.
storage temp.
−20°C
SMILES string
CC(C)CC(NC(=O)C(CO)NC(=O)CNC(=O)C(N)Cc1ccc(O)cc1)C(=O)N2CCCC2C(=O)NC(CCC(N)=O)C(=O)NC(CCCCN)C(=O)NC(CO)C(=O)NC(CCC(N)=O)C(=O)NC(CCCNC(N)=N)C(=O)NC(CO)C(=O)NC(CCC(N)=O)C(=O)NC(CC(O)=O)C(=O)NC(CCC(O)=O)C(=O)NC(CC(N)=O)C(O)=O
InChI
1S/C71H113N23O28/c1-33(2)25-44(90-65(116)46(30-95)82-54(103)29-81-57(108)36(73)26-34-10-12-35(98)13-11-34)69(120)94-24-6-9-49(94)68(119)88-41(16-20-52(76)101)61(112)83-37(7-3-4-22-72)58(109)92-47(31-96)66(117)86-39(14-18-50(74)99)60(111)84-38(8-5-23-80-71(78)79)59(110)93-48(32-97)67(118)87-40(15-19-51(75)100)62(113)89-43(28-56(106)107)64(115)85-42(17-21-55(104)105)63(114)91-45(70(121)122)27-53(77)102/h10-13,33,36-49,95-98H,3-9,14-32,72-73H2,1-2H3,(H2,74,99)(H2,75,100)(H2,76,101)(H2,77,102)(H,81,108)(H,82,103)(H,83,112)(H,84,111)(H,85,115)(H,86,117)(H,87,118)(H,88,119)(H,89,113)(H,90,116)(H,91,114)(H,92,109)(H,93,110)(H,104,105)(H,106,107)(H,121,122)(H4,78,79,80)
InChI key
ROVJYNCPHLNVSO-UHFFFAOYSA-N
Gene Information
guinea pig ... MBP(100731253)
Looking for similar products? Visit Product Comparison Guide
Amino Acid Sequence
Biochem/physiol Actions
Linkage
Other Notes
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service