M5154
Moxisylyte hydrochloride
>99% (TLC), powder
Synonym(s):
4-(2-Dimethylaminoethoxy)-5-isopropyl-2-methylphenyl acetate hydrochloride
About This Item
assay
>99% (TLC)
form
powder
color
off-white
solubility
ethanol: 50 mg/mL
SMILES string
Cl.CC(C)c1cc(OC(C)=O)c(C)cc1OCCN(C)C
InChI
1S/C16H25NO3.ClH/c1-11(2)14-10-15(20-13(4)18)12(3)9-16(14)19-8-7-17(5)6;/h9-11H,7-8H2,1-6H3;1H
InChI key
IPWGSXZCDPTDEH-UHFFFAOYSA-N
Gene Information
human ... ADRA1A(148) , ADRA1B(147) , ADRA1D(146) , ADRA2A(150) , ADRA2B(151) , ADRA2C(152)
Application
- as an α-adrenergic antagonist to study its effects on the circulating level of vitellogenin Ao1 (VtgAo1) or its expression in the liver, heart, and brain of zebrafish [1]
- in a study to examine its binding affinity to human serum albumin (HSA) for predicting plasma protein binding of drugs[2]
- as α1- and α2-adrenoceptor dual antagonist to investigate its effects on antigen-induced airway reactions in guinea pigs[3]
Biochem/physiol Actions
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service