Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M5074

Sigma-Aldrich

Myosin Heavy Chain from human heart

Synonym(s):

Myosin from human heart, MIHC Kinase, Myosin I heavy chain, NHCK

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human heart

assay

≥85%

form

lyophilized

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... MYH1(4619)

Looking for similar products? Visit Product Comparison Guide

Physical form

Lyophilized from salt buffer

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service