Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M4818

Sigma-Aldrich

Anti-phospho-MEK7 (pSer271/pThr275) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

western blot: 0.1-1.0 μg/mL using extracts prepared GST-tagged fusion protein expressing MEK 7.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MAP2K7(5609)
mouse ... Map2k7(26400)

Immunogen

synthetic MEK7-derived phosphopeptide that is double phosphorylated at serine 271 and threonine 275.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, with 1.0 mg/mL BSA (IgG and protease free) and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service