Key Documents
M4784
Sigma-Aldrich
Specimen mounts
For JEOL Instruments
About This Item
UNSPSC Code:
41100000
NACRES:
MA.01
material
aluminum (with lathe finish)
diam. × H
9.5 mm × 9.5 mm
General description
Surface has barely visible rings which help to locate the center.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Customers Also Viewed
Slide 1 of 1