Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M4442

Sigma-Aldrich

Anti-Matrix Metalloproteinase-15, Hinge Region antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MMP-15, Anti-MT2-MMP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP15(4324)

Specificity

Recognizes native and reduced MMP-15. By immunoblotting against the reduced protein, the antibody identifies bands at 72 kDa and 55 kDa, as well as activation/breakdown products at 48 kDa. Often MMP-15 is fully activated and the 48 kDa band is the dominant band seen. Upon activation, MMP-15 may be shed into the culture media and it is unclear if the shed form is proteolytically active.

Immunogen

synthetic peptide corresponding to the hinge region of human matrix metalloproteinase-15.

Physical form

Solution in 0.01 M phosphate buffered saline containing 50% glycerol and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service