Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M4321

Sigma-Aldrich

MYO3β (1-326), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

mysoin lllB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

72-98 nmol/min·mg

mol wt

~63 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MYO3B(140469)

Biochem/physiol Actions

MYO3β is a member of the Class III myosins that are actin-dependent motor proteins containing an amino-terminal kinase domain. MYO3β contains an N-terminal kinase domain, followed by motor, neck, and tail domains. The MYO3β gene generates a variety of splice variants that contain 1 or 2 calmodulin-binding (IQ) motifs in the neck domain and 1 of 3 domains in the tail domain. Northern blot analysis shows expression of a 7-kb MYO3β transcript in the human retina but not in a RPE cell line. The MYO3β gene transcript is also detected in the kidney, intestine and testis. The MYO3β gene maps to chromosome 2q31.1-q31.2 by genomic sequence alignment.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andréa C Dosé et al.
Genomics, 79(5), 621-624 (2002-05-07)
Class III myosins are actin-based motors with amino-terminal kinase domains. Expression of these motors is highly enhanced in retinal photoreceptors. As mutations in the gene encoding NINAC, a Drosophila melanogaster class III myosin, cause retinal degeneration, human homologs of this

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service