Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M4316

Sigma-Aldrich

Anti-Matrix Metalloproteinase-22, N-Terminal antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CMMP, Anti-MMP-22

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP23B(8510)

Specificity

By immunoblotting against the reduced protein, the antibody identifies bands at 53 kDa, as well as larger bands at 110 kDa and 80 kDa. The larger bands are greater than the predicted 53 kDa, which might represent alternative splicing, post-translational modifications, aggregation or other phenomenon.

Immunogen

synthetic peptide corresponding to the N-terminal of active chicken matrix metalloproteinase-22.

Physical form

Solution in 0.01 M phosphate buffered saline contining 50% glycerol and 0.05% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service