Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M4302

Sigma-Aldrich

Anti-Matrix Metalloproteinase-1, C-Terminal antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MMP-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

western blot: 1:1,000 using a concentrated cell culture medium from a stimulated human cell line (M2928)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP1(4312)

Specificity

By immunoblotting against the reduced protein, the antibody reacts with bands at 53 kDa and 51 kDa (the pro-form), as well as active forms. Also reacts with non-reduced MMP-1. Since the hemopexin domain is cleaved off to form the smallest active form of MMP-1, the antibody does not recognize the 21 kDa form of MMP-1.

Immunogen

synthetic peptide corresponding to C-terminal human matrix metalloproteinase-1 (interstitial collagenase).

Physical form

Solution in phosphate buffered saline, pH 7.4, containing 50% glycerol and 15 mM sodium azide

Not finding the right product?  

Try our Product Selector Tool.

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service