Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M4262

Sigma-Aldrich

3′-O-Methyluridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14N2O6
CAS Number:
Molecular Weight:
258.23
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

COC1C(O)C(OC1CO)N2C=CC(=O)NC2=O

InChI

1S/C10H14N2O6/c1-17-8-5(4-13)18-9(7(8)15)12-3-2-6(14)11-10(12)16/h2-3,5,7-9,13,15H,4H2,1H3,(H,11,14,16)

InChI key

YKNATSNMFLEFRB-UHFFFAOYSA-N

Application

3′-O-Methyluridine may be used in comparative studies on the methylation of 2′ and 3′ hydroxyl groups of uridines in plant microRNAs.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingchun Zhang et al.
Journal of the American Society for Mass Spectrometry, 17(8), 1096-1099 (2006-06-06)
Recent studies revealed that the 3'-terminal nucleotides in plant microRNAs were methylated on the ribose at the 2' or 3' hydroxyl groups. Here we examined the fragmentation of the electrospray-produced [M + H]+ and [M - H]- ions of 2'-

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service