M4070
Mu-Phe-hPhe-FMK
≥98% (TLC), solid
Synonym(s):
N-Morpholineurea-phenylalanyl-homophenylalanylfluoromethyl ketone
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C25H30O4N3F
Molecular Weight:
455.52
MDL number:
UNSPSC Code:
12352200
assay
≥98% (TLC)
form
solid
storage condition
desiccated
solubility
DMSO: soluble
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Cathepsin B and L inhibitor peptide
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Anne-Louise Gannon et al.
Frontiers in endocrinology, 11, 599869-599869 (2021-02-16)
Adrenal androgens are fundamental mediators of ovarian folliculogenesis, embryonic implantation, and breast development. Although adrenal androgen function in target tissues are well characterized, there is little research covering the role of androgen-signaling within the adrenal itself. Adrenal glands express AR
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service