Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M4065

Sigma-Aldrich

Anti-Matrix Metalloproteinase-2 antibody produced in rabbit

~1 mg/mL, IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-MMP-2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 72 kDa (proform)

species reactivity

mouse, bovine, rat, human

concentration

~1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 10-20 μg/mL
western blot: 5-10 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMP2(4313)

Specificity

Recognizes MMP-2, cytoplasmic localization. Shows no cross-reactivity with the pro and active forms of other MMPs.

Immunogen

synthetic peptide from the second half of human MMP-2 (Gelatinase-A, 72 kDa Gelatinase). The immunogen is synthesized as a 631 amino acid proenzyme that is activated by cleavage of the the first 80 amino acids.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Solution in 10 mM phosphate buffered saline, pH 7.4, containing 0.2% bovine serum albumin and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andreea Iren Serban et al.
PloS one, 11(3), e0152376-e0152376 (2016-03-26)
AGEs accumulation in the skin affects extracellular matrix (ECM) turnover and triggers diabetes associated skin conditions and accelerated skin aging. The receptor of AGEs (RAGE) has an essential contribution to cellular dysfunction driven by chronic inflammatory responses while TGF-β1 is
Manikantan S Kiran et al.
Vascular cell, 3(1), 6-6 (2011-02-26)
The mechanism of cell-cell contact dependent regulation of pericellular proteolysis in angiogenesis was examined by studying the expression of MMPs using isolated HUVECs in culture. Zymography, Immunoblot and RT-PCR analysis showed that the production and secretion of matrixmetalloproteinase-2 and matrixmetalloproteinase-9

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service