Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M3441

Sigma-Aldrich

Midkine, human

≥90% (SDS-PAGE)

Synonym(s):

Heparin-binding growth/differentiation factor

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

synthetic

Quality Level

assay

≥90% (SDS-PAGE)

packaging

pkg of 50 μg

color

white

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... MDK(4192)

Biochem/physiol Actions

Midkine, a heparin-binding growth factor, contains 121 amino acid residues with 5 disulfide bonds. It promotes the growth, survival, and migration of various cells, and plays roles in neurogenesis and epithelial mesenchymal interactions during organogenesis. The C-terminal portion contains the heparin-binding domain and the antigenicity and neurite-promoting sequences. Both the C- and N-terminals are necessary for the molecule′s neurotrophic effects.
Promotes the growth, survival, and migration of various cells, and plays roles in neurogenesis and epithelial mesenchymal interactions during organogenesis. The C-terminal portion contains the heparin-binding domain and the antigenicity and neurite-promoting sequences. Both the C- and N-terminals are necessary for the molecule′s neurotrophic effects.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service