Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M3196

Sigma-Aldrich

5-MEO-DIPT hydrochloride hydrate

Synonym(s):

Methoxy Foxy, N,N-Diisopropyl-5-methoxytryptamine hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H26N2O · xHCl · yH2O
Molecular Weight:
274.40 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

drug control

USDEA Schedule I

storage temp.

room temp

SMILES string

O.Cl.COc1ccc2[nH]cc(CCN(C(C)C)C(C)C)c2c1

InChI

1S/C17H26N2O.ClH.H2O/c1-12(2)19(13(3)4)9-8-14-11-18-17-7-6-15(20-5)10-16(14)17;;/h6-7,10-13,18H,8-9H2,1-5H3;1H;1H2

InChI key

DFERASOFTGVIPG-UHFFFAOYSA-N

Biochem/physiol Actions

5-MeO-DIPT is a tryptamine derivative. In animal behavioral studies, 5-MeO-DIPT has been shown to produce behavioral effects that are substantially similar to those of LSD, both schedule I hallucinogens.
a synthetic chemical tryptamine structurally related to other tryptamines (natural and synthetic), such as 5-MeO-DMT, N,N-DMT, psiloc(yb) in and serotonin.

Features and Benefits

This compound is featured on the Biogenic Amine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to serotonin receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service