Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

M3043

Sigma-Aldrich

2-Methoxy-4-(2-nitrovinyl)phenyl β-D-galactopyranoside

Synonym(s):

MNP-Gal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H19NO9
CAS Number:
Molecular Weight:
357.31
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

storage temp.

−20°C

SMILES string

COc1cc(ccc1OC2OC(CO)C(O)C(O)C2O)\C=C/N(=O)=O

InChI

1S/C15H19NO9/c1-23-10-6-8(4-5-16(21)22)2-3-9(10)24-15-14(20)13(19)12(18)11(7-17)25-15/h2-6,11-15,17-20H,7H2,1H3/b5-4-

InChI key

VLHYYNTWHBRKNT-PLNGDYQASA-N

General description

2-Methoxy-4-(2-nitrovinyl)phenyl β-D-galactopyranoside is a substrate for β-galactosidase.

Application

2-Methoxy-4-(2-nitrovinyl)phenyl β-D-galactopyranoside can be used as a substrate for the assay of NAG and β-D-galactosidase. It has also been used in a study to evaluate new chromogenic substrates for the detection of coliforms.

Substrates

Chromogenic substrate (505 nm) for β-galactosidase

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C T Yuen et al.
Clinica chimica acta; international journal of clinical chemistry, 124(2), 195-204 (1982-09-15)
(1) The synthesis of 2-methoxy-4-(2'-nitrovinyl)-phenyl 2-acetamido-2-deoxy-beta-D-glucopyranoside (MNP-GlcNAc) and 2-methoxy-4-(2'nitrovinyl)-phenyl beta-D-galactopyranoside (MNP-Gal) as substrates for the assay of NAG and beta-d-galactosidase are described. (2) beta-Glycosidase activities were determined in random urine samples from normal males and females aged between 12 and
N K Browne et al.
Journal of applied microbiology, 108(5), 1828-1838 (2009-11-03)
To evaluate a new range of chromogenic substrates for the detection of beta-galactosidase activity in coliforms and to compare their performance in agar media and broths. Sixteen novel galactoside substrates were prepared and incorporated into agar and broth. Their performance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service