Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M2787

Sigma-Aldrich

2-Oleoylglycerol

≥94% (TLC)

Synonym(s):

MG(0:0/18:1(9Z)/0:0), 2-Monoolein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H40O4
CAS Number:
Molecular Weight:
356.54
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

Quality Level

assay

≥94% (TLC)

form

liquid

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[H]C(CO)(OC(CCCCCCC/C=C\CCCCCCCC)=O)CO

InChI

1S/C21H40O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-20(18-22)19-23/h9-10,20,22-23H,2-8,11-19H2,1H3/b10-9-

InChI key

UPWGQKDVAURUGE-KTKRTIGZSA-N

Biochem/physiol Actions

2-Monoolein (2-Oleoylglycerol) is used as a GPR119 agonist than regulates GLP-1 release. 2-Monoolein may be used to differentiate and study the kinetics of monoacylglycerol lipase(s).

Packaging

Packaged under Argon.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dong Cheng et al.
The Journal of biological chemistry, 283(44), 29802-29811 (2008-09-05)
Acyl coenzyme A:diacylglycerol acyltransferase 1 (DGAT1) is one of the four intestinal membrane bound acyltransferases implicated in dietary fat absorption. Recently, it was found that, in addition to acylating diacylglycerol (DAG), DGAT1 also possesses robust enzymatic activity for acylating monoacylglycerol
Katrine B Hansen et al.
The Journal of clinical endocrinology and metabolism, 96(9), E1409-E1417 (2011-07-23)
Dietary fat is thought to stimulate release of incretin hormones via activation of fatty acid receptors in the intestine. However, dietary fat (triacylglycerol) is digested to 2-monoacylglycerol and fatty acids. Activation of G protein-coupled receptor 119 (GPR119) stimulates glucagon-like peptide-1
Christopher J Fowler et al.
Biochemical pharmacology, 69(8), 1241-1245 (2005-03-30)
In the present study, the abilities of cyclooxygenated derivatives of 1-arachidonoylglycerol and related compounds to prevent the metabolism of [3H]2-oleoylglycerol and [3H]anandamide by cytosolic and membrane fractions, respectively, have been investigated. For each compound, nine concentrations (range 0.2-100 microM) were
S Vandevoorde et al.
British journal of pharmacology, 150(2), 186-191 (2006-12-05)
Two compounds, URB602 and URB754, have been reported in the literature to be selective inhibitors of monoacylglycerol lipase, although a recent study has questioned their ability to prevent 2-arachidonoyl hydrolysis by brain homogenates and cerebellar membranes. In the present study
Verena Hiebl et al.
Biological procedures online, 22, 7-7 (2020-04-21)
The human Caco-2 cell line is a common in vitro model of the intestinal epithelial barrier. As the intestine is a major interface in cholesterol turnover and represents a non-biliary pathway for cholesterol excretion, Caco-2 cells are also a valuable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service