Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M2570

Sigma-Aldrich

L-Methionine γ-Lyase

≥0.2 unit/mg solid

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204

recombinant

expressed in E. coli

form

lyophilized powder

specific activity

≥0.2 unit/mg solid

storage temp.

−20°C

General description

L-Methionine γ-lyase is a pyridoxal phosphate-containing enzyme that converts L-methionine to α-ketobutyrate, ammonia and methyl mercaptan. [1]

Application

L-Methionine γ-lyase has been used in a study to assess catabolism of volatile sulfur compound precursors by Brevibacterium linens and Geotrichum candidum. [2] It has also been used in a study to investigate the enzymatic processing of fluorinated methionine analogs. [1]

Biochem/physiol Actions

L-Methionine γ-Lyase or methioninase has been shown to have anti-tumor effects by depleting methionine from methionine-dependent tumors making them more sensitive to traditional chemotherapies. [3]

Unit Definition

One unit iwill convert 1 micromole of L-methionine to 2-ketobutyrate per minute at pH 7.0 at 37 °C.

Physical form

Supplied as a lyophilized powder containing pyridoxal-5′-phosphate and potassium phosphate

Preparation Note

Expressed in E. coli from a proprietary gene

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A S El-Sayed et al.
Journal of applied microbiology, 111(1), 54-69 (2011-04-07)
To immobilize the purified Aspergillus flavipesl-methioninase on solid carriers for continuous production of methanethiol with high purity, by the enzymatic methods. The purified l-methioninase was immobilized using different methods, and physicochemical and kinetic studies for the potent immobilized enzyme were
[Perspectives of developing enzyme-based antitumor drugs].
V S Pokrovskiĭ et al.
Voprosy onkologii, 57(2), 155-164 (2011-08-04)
Ashraf S A El-Sayed
Journal of microbiology (Seoul, Korea), 49(1), 130-140 (2011-03-04)
L-Methioninase was purified to electrophoretic homogeneity from cultures of Aspergillus flavipes using anion-exchange and gel filtration chromatography by 12.1 fold compared to the crude enzyme preparation. The purified enzyme had a molecular mass of 47 kDa under denaturing conditions and
Microdetermination of D-amino acids and D-amino acid oxidase activity with 3,methyl-2-benzothiazolone hydrazone hydrochloride.
K Soda
Analytical biochemistry, 25(1), 228-235 (1968-10-24)
Xinghua Sun et al.
Cancer research, 63(23), 8377-8383 (2003-12-18)
Recombinant methioninase (rMETase) is an enzyme active in preclinical mouse models of human cancer. The efficacy of rMETase is due to depletion of plasma methionine, an amino acid for which tumors generally have an abnormally high methionine requirement. Furthermore, transient

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service