Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M208

Sigma-Aldrich

Monocyte Chemotactic Protein-1 from rat

>97% (SDS-PAGE), recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture

Synonym(s):

MCP-1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800

biological source

rat

recombinant

expressed in E. coli

assay

>97% (SDS-PAGE)

form

lyophilized powder

potency

100 ng/mL ED50

mol wt

14.1 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles (Storage in "frost-free" freezers is not recommended)

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

rat ... Ccl2(24770)

Physical form

Lyophilized from a 0.2 μm filtered solution

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service