Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M143130

Sigma-Aldrich

6-Hexachloro-Fluorescein-Azide

Synonym(s):

3-Azidopropylaminocarbonyl-2′,4,4′,5′,7,7′-hexachlorofluorescein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H12Cl6N4O6
Molecular Weight:
665.09
UNSPSC Code:
12352200
NACRES:
NA.21

product line

Proligo Reagents

assay

≥95.0% (reversed phase HPLC)

form

solid

color

orange to red

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR

storage temp.

−20°C

SMILES string

OC1=C(Cl)C2=C(C=C1Cl)C(OC3=O)(C4=C3C(Cl)=CC(C(NCCCN=[N+]=[N-])=O)=C4Cl)C5=CC(Cl)=C(O)C(Cl)=C5O2

InChI

1S/C24H12Cl6N4O6/c25-10-4-7(22(37)32-2-1-3-33-34-31)15(28)14-13(10)23(38)40-24(14)8-5-11(26)18(35)16(29)20(8)39-21-9(24)6-12(27)19(36)17(21)30/h4-6,35-36H,1-3H2,(H,32,37)

InChI key

HGKPJGGICCDTJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service