Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M1194

Sigma-Aldrich

Anti-MTA2/MTA1L1 (RT-16) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Metastasis-associated Gene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 73 kDa

species reactivity

human, rat, mouse

technique(s)

immunoprecipitation (IP): 1-2 μL using 293-T cell lysate
indirect immunofluorescence: 1:100-1:200 using methanol-acetone fixed C2 myoblasts
western blot: 1:2,000-1:4,000 using nuclear extracts of MCF7 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTA2(9219)
mouse ... Mta2(23942)
rat ... Mta2(361724)

General description

Metastasis-associated protein (MTA2) comprises bromo-adjacent homology (BAH), Egl-27 and MTA1 homology 2 (ELM2), Swi3, Ada2, N-Cor, and TFIIIB (SANT), and an atypical GATA zinc finger domain. It is a phosphorylated protein. MTA2 gene is mapped to human chromosome 11q12.3. Human MTA2 shares approximately 80% overall homology to human MTA1 and MTA3 proteins, with the C-terminus being more divergent than the N-terminus.

Specificity

Anti MTA2/MTA1L1 (RT-16) specifically recognizes MTA2/MTA1L1.

Immunogen

synthetic peptide corresponding to amino acids 626-641 of human MTA2/MTA1L1, conjugated to KLH via an N-terminal added cysteine residue. The immunizing peptide is conserved in human, mouse, and rat, and is not present in other members of the family.

Application

Anti-MTA2/MTA1L1 (RT-16) antibody produced in rabbit has been used in:
  • tandem immunoprecipitation[1]
  • immunoblotting
  • immunofluorescence

Biochem/physiol Actions

Metastasis-associated protein (MTA2) mediates cytoskeletal reorganization. It interacts with the histone deacetylases (HDAC) within the nuclear remodeling and deacetylation complexes namely, nucleosome remodeling and deacetylase (Mi2/NuRD). This interaction mediates transcriptional repression. Also called p53 target protein in deacetylase complex (PID), it regulates p53 function and represses p53-dependent transcriptional activation, growth arrest and apoptosis. MTA2 also favors tumor growth and migration.

Physical form

Solution in 0.01 M phophate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. Storage in frost-free freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Weijun Ding et al.
International journal of clinical and experimental pathology, 8(6), 7173-7180 (2015-08-12)
Association of MTA2 expression with presence, development, metastasis and prognosis of colorectal cancer (CRC) was investigated. 90 CRC-related cases with follow-up information were made into tissue microarrays according to the paired principle of cancer tissues and the adjacent tissues. Subsequently
Role of MTA2 in human cancer
Covington KR and Fuqua SAW
Cancer Metastasis Reviews, 921-928 (2014)
TP53 mutation, epithelial-mesenchymal transition, and stemlike features in breast cancer subtypes
Coradini D, et al.
Journal of Biomedicine and Biotechnology (2012)
J Luo et al.
Nature, 408(6810), 377-381 (2000-12-01)
The p53 tumour suppressor is a transcriptional factor whose activity is modulated by protein stability and post-translational modifications including acetylation. The mechanism by which acetylated p53 is maintained in vivo remains unclear. Here we show that the deacetylation of p53
Gohei Nishibuchi et al.
The Journal of biological chemistry, 289(42), 28956-28970 (2014-09-06)
Histone H3K4 methylation has been linked to transcriptional activation. KDM5A (also known as RBP2 or JARID1A), a member of the KDM5 protein family, is an H3K4 demethylase, previously implicated in the regulation of transcription and differentiation. Here, we show that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service