Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M0945

Sigma-Aldrich

Anti-Maspin antibody produced in rabbit

~0.5 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-PI5, Anti-Protease Inhibitor 5, Anti-SERPINB5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

concentration

~0.5 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 10-20 μg/mL using heat retrieved, human prostate sections
western blot: 1-2 μg/mL using human A431 cell extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SERPINB5(5268)

General description

Maspin gene is located on chromosome 18q21.3 in a cluster of genes encoding members of the ovalbumin family of serine protease inhibitors. Expression of maspin was found also in human cornea in the epithelium, endothelium and stroma cells.

Immunogen

synthetic peptide corresponding to amino acids 333-348 of human maspin with a C-terminal added cysteine, conjugated to KLH.

Application

Anti-Maspin antibody produced in rabbit has been used in immunoblotting and immunohistochemistry.

Biochem/physiol Actions

Maspin is a serine protease inhibitor (serpin) with tumor suppressive activities. Maspin protein was identified in a search for tumor suppressor genes that are defective in human breast carcinoma cells. Maspin is expressed in normal mammary epithelial cell strains, but not in most mammary tumor cell lines. Re-expression of maspin in mammary carcinoma cells does not alter the growth properties of the cells in vitro, but reduces their ability to induce tumors, to metastasize in nude mice, and to invade a basement membrane matrix in vitro. The molecular mechanism of maspin action is not clear. Using a two-hybrid system, it has been found that heat shock protein 90, glutathione S-transferase (GST), and heat shock protein 70 interacted with maspin with the highest frequencies. This protein exerts its tumor suppressive role, at least in part, by blocking the activity of pericellular uPA (urokinase Plasminogen Activator).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide and 1% inactivated bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maspin-a novel protease inhibitor with tumor-suppressing activity in breast cancer
Maass N, et al.
Acta Oncologica (Stockholm, Sweden), 39(8), 931-934 (2000)
Maspin expression inhibits osteolysis, tumor growth, and angiogenesis in a model of prostate cancer bone metastasis
Cher ML, et al.
Proceedings of the National Academy of Sciences of the USA, 100(13), 7847-7852 (2003)
Maspin is not required for embryonic development or tumour suppression
Teoh SSY, et al.
Nature Communications, 5, 3164-3164 (2014)
Tumor-suppressive maspin regulates cell response to oxidative stress by direct interaction with glutathione S-transferase
Yin S, et al.
The Journal of biological chemistry, 280(41), 34985-34996 (2005)
Maspin: synthesis by human cornea and regulation of in vitro stromal cell adhesion to extracellular matrix
Ngamkitidechakul C, et al.
Investigative Ophthalmology & Visual Science, 42(13), 3135-3141 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service