Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M0820

Sigma-Aldrich

Anti-MOG (C-terminal) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Myelin-oligodendrocyte Glycoprotein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 26 kDa

species reactivity

rat

technique(s)

western blot: 2.5-5 μg/mL using rat brain homogenate (S1 fraction) and rat spinal cord homogenate (P2 fraction)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... MOG(4340)
rat ... Mog(24558)

General description

Myelin/oligodendrocyte glycoprotein (MOG) is an integral membrane glycoprotein belonging to the large immunoglobulin superfamily. It is a 26-28kDa glycoprotein localized on chromosome 6p21.3. It consists of an extracellular immunoglobulin-like domain and two hydrophobic regions. It is expressed on the surface of myelinating oligodendrocytes and external lamellae of myelin sheaths in the central nervous system.

Immunogen

synthetic peptide corresponding to amino acids 231-247 located near the C-terminus of human MOGα1 isoform, conjugated to KLH. This sequence is identical in human MOGα4 and MOGβ1 isoforms, partially found (six amino acids at the C-terminus) in human MOGα2 and MOGα3 isoform, and identical in rat, mouse, and bovine MOG.

Application

Anti-MOG (C-terminal) antibody produced in rabbit is suitable for western blot at a concentration of 2.5-5 ?g/mL using rat brain homogenate (S1 fraction) and rat spinal cord homogenate (P2 fraction).

Biochem/physiol Actions

Myelin/oligodendrocyte glycoprotein (MOG) is a major component of the central nervous system. It is involved in several cellular processes including cellular adhesion, in the regulation of oligodendrocyte microtubule stability and facilitates the interactions between myelin and the immune system. It has been reported that MOG may be involved in the completion and/or maintenance of the myelin sheath, and in cell-cell communication. It has also been observed that mutation or polymorphisms in MOG gene could cause multiple sclerosis.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B della Gaspera et al.
European journal of biochemistry, 258(2), 478-484 (1999-01-05)
Myelin/oligodendrocyte glycoprotein (MOG), a specific component of the mammalian central nervous system, is located on the surface of the oligodendrocyte plasma membrane and the outermost lamellae of mature myelin; it is expressed during the latter steps of myelinogenesis. It has
T G Johns et al.
Journal of neurochemistry, 72(1), 1-9 (1999-01-14)
Myelin oligodendrocyte glycoprotein (MOG) is a quantitatively minor component of CNS myelin whose function remains relatively unknown. As MOG is an autoantigen capable of producing a demyelinating multiple sclerosis-like disease in mice and rats, much of the research directed toward
D Pham-Dinh et al.
Genomics, 29(2), 345-352 (1995-09-20)
Myelin/oligodendrocyte glycoprotein (MOG), a specific component of the central nervous system localized on the outermost lamellae of mature myelin, is a member of the immunoglobulin superfamily. We report here the organization of the human MOG gene, which spans approximately 17
D Pham-Dinh et al.
Proceedings of the National Academy of Sciences of the United States of America, 90(17), 7990-7994 (1993-09-01)
Myelin/oligodendrocyte glycoprotein (MOG) is found on the surface of myelinating oligodendrocytes and external lamellae of myelin sheaths in the central nervous system, and it is a target antigen in experimental autoimmune encephalomyelitis and multiple sclerosis. We have isolated bovine, mouse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service