Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

M0819

Sigma-Aldrich

Anti-MTA3L antibody produced in rabbit

enhanced validation

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Metasisis-associated Gene

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 75 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

microarray: suitable
western blot: 1:1,000-1:2,000 using transfected cell extracts of HEK 293-T cells

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MTA3(57504)

General description

Metastasis associated 1 family member 3 (MTA3) is a component of the Mi-2/ nucleosome remodeling and deacetylase (NURD) complex. It comprises the bromo-adjacent homology (BAH) GATA-zinc finger (ZF), Egl27/MTA1 (ELM), SW13, ADA2, N-CoR and TFIIIB domain (SANT) domains. The MTA3 gene is located on human chromosome 2p21 and encodes for two isoforms (MTA3 and MTA3L).

Specificity

Anti-MTA3L recognizes specifically human MTA3L.

Immunogen

synthetic peptide corresponding to amino acids 551-568 of human MTA3L, conjugated to KLH via an N-terminal added lysine residue. The immunizing sequence is not present in other members of the family, MTA1 and MTA2.

Application

Anti-MTA3L antibody produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

Metastasis associated 1 family member 3 (MTA3) is an estrogen receptor (ER)-regulated component of the Mi-2/ nucleosome remodeling and deacetylase (NURD) complex transcriptional corepressor complex. This complex is crucial for the initiation of hematopoiesis. Post estrogen binding to the estrogen receptor, MTA3, in conjunction with other NuRD components, decreases expression of the transcriptional repressor Snail, ultimately modulating E-cadherin expression and maintenance of normal epithelial architecture. Loss of MTA3 or ER leads to deregulated expression of Snail, transcriptional repression of E-cadherin, and a consequent predisposition to invasive cell growth in breast epithelial cancers. MTA3 plays a role in B cell fate determination, through interaction with the transcription factor B cell lymphoma 6 (BCL-6). It also regulates epithelial-mesenchymal transitions.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, and 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in frost-free freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bramanandam Manavathi et al.
The Journal of biological chemistry, 282(3), 1529-1533 (2006-12-05)
Regulation of fundamental genetic processes demands dynamic participation of transcription factors, their coregulators, and multiprotein chromatin remodeling activities at target genes. One family of chromatin modifiers that is ubiquitously expressed is the metastasis tumor antigens (MTA), which are integral parts
Carissa Dege et al.
Immunological reviews, 261(1), 126-140 (2014-08-16)
Mi-2/nucleosomal remodeling and deacetylase (NuRD) complexes are important epigenetic regulators of chromatin structure and gene expression. Mi-2/NuRD complexes are an assemblage of proteins that combine key epigenetic regulators necessary for (i) histone deacetylation and demethylation, (ii) binding to methylated DNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service