Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M0442

Sigma-Aldrich

Microsomes from Liver, Pooled

from human(CMV-negative)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161501
NACRES:
NA.47

biological source

human (CMV-negative)

Quality Level

form

liquid

packaging

vial of ~10 mg

shipped in

dry ice

storage temp.

−70°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Liver microsomes are subcellular particles derived from the endoplasmic reticulum of hepatic cells. These microsomes are a rich source of drug metabolizing enzymes, including cytochrome P-450. Microsome pools from various sources are useful in the study of xenobiotic metabolism and drug interactions.
Source of drug metabolizing enzymes, including cytochrome P-450.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service