Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M022000

Sigma-Aldrich

TFA-Pentylaminolinker Phosphoramidite

Synonym(s):

TFA-Pentylaminolinker amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H29F3N3O3P
Molecular Weight:
399.39
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
Pricing and availability is not currently available.

type

for DNA synthesis

product line

Proligo Reagents

assay

≥99% (31P-NMR)

suitability

conforms to structure for H-NMR (Identity)

storage temp.

−70°C

SMILES string

CC(C)N(C(C)C)P(OCCCCCNC(=O)C(F)(F)F)OCCC#N

InChI

1S/C16H29F3N3O3P/c1-13(2)22(14(3)4)26(25-12-8-9-20)24-11-7-5-6-10-21-15(23)16(17,18)19/h13-14H,5-8,10-12H2,1-4H3,(H,21,23)

InChI key

OVFVLUKVODOLBV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service