Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M0198

Sigma-Aldrich

MUSK (519-end), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC126323, MGC126324

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

6-8 nmol/min·mg

mol wt

~63 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MUSK(4593)

Biochem/physiol Actions

MUSK is a receptor tyrosine kinase necessary for neuromuscular junction formation.MUSK gene expression is highly regulated during neuromuscular junction and it is involved in intercellular communication present on the surface of cells that are activated by specific protein ligands. MUSK members play a key role in growth and differentiation of those cell types. Agrin signals through MUSK to cluster acetylcholine receptors on the postsynaptic membrane of the neuromuscular junction. DOK7, a MUSK-interacting cytoplasmic protein, is essential for MUSK activation in cultured myotubes. MUSK also plays a critical role in the development of normal blood vessels.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang-Hoon Kim et al.
The Journal of biological chemistry, 278(40), 38522-38527 (2003-07-30)
MuSK is a receptor tyrosine kinase essential for neuromuscular junction formation. Expression of the MuSK gene is tightly regulated during development and at the neuromuscular junction. However, little is known about molecular mechanisms regulating its gene expression. Here we report
Kumiko Okada et al.
Science (New York, N.Y.), 312(5781), 1802-1805 (2006-06-24)
The formation of the neuromuscular synapse requires muscle-specific receptor kinase (MuSK) to orchestrate postsynaptic differentiation, including the clustering of receptors for the neurotransmitter acetylcholine. Upon innervation, neural agrin activates MuSK to establish the postsynaptic apparatus, although agrin-independent formation of neuromuscular

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service