Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M0191

Sigma-Aldrich

DL-p-Methoxymethamphetamine hydrochloride solution

BioReagent, 1.0 mg/mL in methanol, solution

Synonym(s):

DL-PMMA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H17NO · HCl
Molecular Weight:
215.72
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

BioReagent

form

solution

drug control

kontrollierte Droge in Deutschland

concentration

1.0 mg/mL in methanol

SMILES string

Cl[H].CNC(C)Cc1ccc(OC)cc1

InChI

1S/C11H17NO.ClH/c1-9(12-2)8-10-4-6-11(13-3)7-5-10;/h4-7,9,12H,8H2,1-3H3;1H

InChI key

IQZVXWOBOYTPER-UHFFFAOYSA-N

Biochem/physiol Actions

May have MDMA (3,4-methylenedioxyphenylisopropylamine; ectasy) effects.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service