Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

M0162

Supelco

Microbead standards

Microbeads prepared from hydrophobic, polymeric material, dual conjugated to fluorescein isothiocyanate (FITC) and R-Phycoerythrin (PE), buffered aqueous suspension

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

form

buffered aqueous suspension

storage temp.

2-8°C

Specificity

For flow cytometric analysis. Designed to simulate the forward scatter and fluorescent intensity generated by fluorochrome-labeled cells and should imitate the properties of labeled human leukocytes. Use 0.1 mL per test.

Physical form

Suspended in a buffered solution with surfactants and sodium azide as preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service