Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

LO2219

Sigma-Aldrich

Cardiotoxicity Ligand Set

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.77

shipped in

dry ice

storage temp.

−20°C

General description

A set of 52 compounds are arranged in 96-well format, one compound per well (250 μl at 10 mM in DMSO)

Application

Cardiotoxicity Ligand Set has been used in the cumulative drug dosage studies on engineered human induced pluripotent stem cell-derived cardiomyocyte (hiPS-CM) tissues.[1]

Biochem/physiol Actions

Cardiotoxicity is an important cause for concern in drug development and appears to cut across therapeutic indications and drug classes. Several drugs have been removed from the market recently because of prolonging the QT interval or causing Torsades de Pointes (TdP).

The Cardiotoxicity Ligand Set features a wide variety of drugs including anti-arrhythmics, antihistamines, antibiotics, anticonvulsants, antidepressants and antipsychotics that are known to have cardiotoxic side-effects.

Features and Benefits

SDFile provided to construct a database containing:
  • Structure
  • Primary Name
  • Secondary Name
  • Pharmacological Activity
  • Product Number
  • Rack Position
This compound is featured on the Histamine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 1 - Carc. 1A - Lact. - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qi Tang et al.
Antiviral research, 173, 104650-104650 (2019-11-18)
IRES-driven translation plays an essential role in picornavirus infection. However, there are seldom reports of compounds targeting this pathway with effective protection in animal models. Here, we identified emetine, an antiprotozoal drug, which inhibits EV-A71 with an EC50 value of
Drug-induced prolongation of the QT interval.
Dan M Roden
The New England journal of medicine, 350(10), 1013-1022 (2004-03-05)
L Testai et al.
Current medicinal chemistry, 11(20), 2691-2706 (2004-11-17)
Many non-cardiovascular drugs of common clinical use cause, as an unwanted accessory property, the prolongation of the cardiac repolarisation process, due to the block of the HERG (Human Ether-a-go-go Related Gene) potassium channel, responsible for the repolarising I(Kr) current. This
Alexander N Katchman et al.
The Journal of pharmacology and experimental therapeutics, 316(3), 1098-1106 (2005-11-10)
The purpose of the present study was to comparatively evaluate human HERG currents and QT intervals following challenge with suspected torsadogenic and nontorsadogenic drugs. Various concentrations of 14 different drugs were initially evaluated in terms of their relative potency to
Cardiac microphysiological devices with flexible thin-film sensors for higher-throughput drug screening
Lind JU, et al.
Lab on a chip, 17(21), 3692-3703 (2017)

Articles

We offers many products related to histamine receptors for your research needs.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service