Key Documents
LIDF455-EP
Lid for Eppendorf® F-45-32-5-PCR Rotor
not aerosol-tight, for 5424/5424R, aluminum
Select a Size
1 EA
$280.00
$280.00
Available to ship onMay 02, 2025Details
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
Change View
1 EA
$280.00
About This Item
UNSPSC Code:
41103913
$280.00
Available to ship onMay 02, 2025Details
Notify Me
Get notified when this item is ready to ship via email.
material
aluminum
feature
not aerosol-tight
manufacturer/tradename
Eppendorf® 022653148 (NA)
Eppendorf® 5424708005 (EU)
technique(s)
PCR: suitable
centrifugation: suitable
compatibility
for use with 5424 R
for use with 5424
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)