Key Documents
LIDF451817-EP
Lid for Eppendorf(R) FA-45-18-17-Cryo Rotor
not aerosol-tight, for 5430/5430R, aluminum
Select a Size
1 EA
$280.00
$280.00
Check Cart for Availability
Select a Size
Change View
1 EA
$280.00
About This Item
UNSPSC Code:
41103913
$280.00
Check Cart for Availability
material
aluminum
feature
not aerosol-tight
manufacturer/tradename
Eppendorf® 022654187 (NA)
Eppendorf® 5427707000 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with 5430 R
for use with 5430
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)