Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

L9894

Sigma-Aldrich

Lectin from Dolichos biflorus (horse gram)

agarose conjugate, buffered aqueous suspension

Synonym(s):

Dolichos biflorus agglutinin, DBA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

conjugate

agarose conjugate

form

buffered aqueous suspension

extent of labeling

2-4 mg protein (Lowry) per mL

matrix

4% beaded agarose

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

DBA has anti-A1 human blood group specificity, useful for distinguishing between A1 and A2 blood types. DBA has strong anti-Cad activity, and has an affinity for terminal N-acetyl-α-D-galactosaminyl residues. Conjugates are prepared from affinity purified lectin.

Physical form

Suspension in 10 mM HEPES, pH 7.5, 0.15 M NaCl, 0.1 mM CaCl2, 20 mM GalNAc, 0.08% sodium azide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service